Learn More
BINDER™ CELLROLL Set
CELLROLL Set is a modular, expandable roller bottle system for cell cultivation.
8080.00€ - 9465.00€
Specifications
For Use With (Equipment) | BINDER Series CB and CBF |
---|---|
Material | Stainless Steel |
Type | Cell Roller |
For Use With (Application) | Cell Cultivation |
Product Code | Brand | Model | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Quantity | Price | Quantity & Availability | |||||
17313683
|
BINDER™
8612-0023 |
CBF 170 | 4 Roller Bottles/Pk. |
8080.00€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
17323683
|
BINDER™
8612-0024 |
CBF 260 | 6 Roller Bottles/Pk. |
9465.00€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Adjustable direction of rotation, speed of rotation, pause times, and interval cycles
- All drive rollers for the bottles run without vibrations on special ball bearings
- Driven by a stepper motor with a soft start and stop phase
- Control unit is outside the incubator and is connected to the interior motor unit via an electric access port
Specifications
BINDER Series CB and CBF | |
Stainless Steel | |
Cell Roller | |
Cell Cultivation |
Product Title
BINDER™ CELLROLL Set