Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Greiner Bio-One 781165
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic Non-cytotoxicCompatible with automated systemsFlexible working volumeRounded square well profile
- Stackable
- Alphanumeric well cod
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
15 to 110 μL | |
Cell Culture | |
Rounded Square | |
10 |
Without Lid | |
Clear | |
384 | |
Sterile | |
131 μL | |
40 |