Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Color:White; Sterility:Sterile; Lid:With Lid
Brand: Greiner Bio-One 675083
Description
Alphanumeric coding
Specifications
Half-Area | |
0.15 cm2 | |
8 x 12 | |
White | |
96 | |
Sterile | |
199 μL | |
Yes | |
32 | |
32/Cs. | |
White |
With Lid | |
Luminescence | |
Not Autoclavable | |
Polystyrene | |
15 to 175 μL | |
Cell Culture | |
Round, Flat Bottom (F-well) | |
4800 x g (swinging bucket rotor) | |
8 | |
Yes | |
127.76 x 85.48 x 14.40 mm |