Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Color:White; Sterility:Sterile; Lid:With Lid
Brand:  Greiner Bio-One 675083
Product Code. 10276893
  • 260.44€ / Pack of 32
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Alphanumeric coding
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Half-Area
0.15 cm2
8 x 12
White
96
Sterile
199 μL
Yes
32
32/Cs.
White
With Lid
Luminescence
Not Autoclavable
Polystyrene
15 to 175 μL
Cell Culture
Round, Flat Bottom (F-well)
4800 x g (swinging bucket rotor)
8
Yes
127.76 x 85.48 x 14.40 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate > White