Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand:  Greiner Bio-One 675180
Product Code. 10288283
  • 89.04€ / Pack of 32
Estimated Shipment: 13-05-2025
Description
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Half-Area
0.15 cm2
Not Autoclavable
Polystyrene
15 to 175 μL
Cell Culture
Round, Flat Bottom (F-well)
4800 x g (swinging bucket rotor)
8
32/Cs.
Clear
With Lid
8 x 12
Clear
96
Sterile
199 μL
Yes
32
Standard
Yes
127.76 x 85.48 x 14.40 mm
Product Title
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate > Clear