missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Color:Black; Sterility:Sterile; Lid:With Lid

Brand:  Greiner Bio-One 675086

Product Code. 10463275

  • 260.44€ / Pack of 32
Only null left
Estimated Shipment: 24-06-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Alphanumeric coding
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Half-Area
Fluorescence
Not Autoclavable
Polystyrene
15 to 175 ÎĽL
Cell Culture
Round, Flat Bottom (F-well)
4800 x g (swinging bucket rotor)
8
Yes
Chimney
With Lid
8 x 12
Black
96
Sterile
199 ÎĽL
Yes
32
32/Cs.
Black
127.76 x 85.48 x 14.40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate > Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.