Learn More
Greiner Bio-One CELLSTAR High-Profile Cell Culture Flasks
Free of detectable DNase, RNase and human DNA
309.00€
Specifications
Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic |
---|---|
Closure Material | HDPE |
Capacity (Metric) | 650 mL |
Material | Polystyrene |
Neck Style | Canted |
Description
- Design of flasks allows efficient access to the cell lawn with cell scrapers and optimal access with pipettes
- Graduation on the side
- Neck of the high profile flask reduces risk of wetting inside of cap with medium, and thus provides additional protection against contamination
- Screw cap
Specifications
Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic | |
650 mL | |
Canted | |
4 | |
Sterile |
HDPE | |
Polystyrene | |
40 | |
Yes |