Learn More
Greiner Bio-One CELLSTAR Lo-Profile Suspension Culture Flask
Ideal for suspension cultures, hybridoma and embryonic stem cells. Greiner Bio-One CELLSTAR Lo-Profile Suspension Culture Flask is free of detectable DNase, RNase and human DNA.
Brand: Greiner Bio-One 660190
This item is not returnable.
View return policy
Description
- Graduation on both sides
- Canted neck style
- Plug seal closure
Specifications
White | |
Canted | |
Plug |
Lo-profile | |
Sterile | |
550 mL |