Learn More
Greiner Bio-One CELLSTAR Lo-Profile Suspension Culture Flask
Ideal for suspension cultures, hybridoma and embryonic stem cells. Greiner Bio-One CELLSTAR Lo-Profile Suspension Culture Flask is free of detectable DNase, RNase and human DNA.
Brand:  Greiner Bio-One 660190
Product Code. 10244372
  • 340.00€ / Pack of 50
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Graduation on both sides
  • Canted neck style
  • Plug seal closure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Canted
Plug
Lo-profile
Sterile
550 mL