Learn More
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks
Designed to culture cell suspended in a growth medium. Greiner Bio-One™ CELLSTAR Suspension Cell Culture Flasks are manufactured from polystyrene for optimum visibility and strength.
Brand: Greiner Bio-One 690190
This item is not returnable.
View return policy
Description
- Hydrophobic surface, ideal for suspension cultures and hybridoma
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic
- Graduation on the side
- Canted neck
- Easily distinguished by their white screw caps
- White standard screw cap with ventilation position
- TC surface treatment: -/suspension
- Sterile and user friendly packaging
Specifications
White | |
Polystyrene | |
200 | |
Screw |
50 mL | |
200 Pack | |
Yes |
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks > Screw Cap Type: White Standard Screw Cap; Capacity: 50mL