Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
97.20€ - 145.00€
Specifications
Quantity | 25 Pk. |
---|---|
Material | Cellulose |
For Use With (Application) | Analysis of soil contaminants, Analysis of soil contaminants |
Product Type | Extraction Thimbles |
Product Code | Brand | Diameter (Metric) Inside | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Height (Metric) | Price | Quantity & Availability | |||||
10620303
|
Macherey-Nagel™
645006 |
22 mm | 80 mm |
97.20€
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10022751
|
Macherey-Nagel™
645024 |
31 mm | 118 mm |
145.00€
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10422032
|
Macherey-Nagel™
645025 |
31 mm | 180 mm |
145.00€
Pack of 25 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
|||||
10536101
|
Macherey-Nagel™
645004 |
15 mm | 100 mm |
97.20€
Pack of 25 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
Description
- Manufactured from pure cellulose
- Used in the fields of air and waste gas analysis for collecting solid particles
- The choice of analytical chemists using Soxhlet or automated extractors
Specifications
25 Pk. | |
Cellulose | |
Analysis of soil contaminants, Analysis of soil contaminants | |
Extraction Thimbles |
Product Title
Macherey-Nagel™ Cellulose Extraction Thimbles