Learn More
Sartorius Cellulose Extraction Thimbles
GREENER_CHOICE
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
Brand:  Sartorius FT-1201-035150
189.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30052907
  • 208.00€ / Pack of 25
Estimated Shipment: 23-07-2025
Description
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
35 mm
1.7 mm
150 mm
Cellulose
C300
Extraction Thimbles
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius Cellulose Extraction Thimbles > Inner Diameter: 35 mm; Length: 150 mm; Wall Thickness: 1.7 mm