Learn More
Velp Scientifica™ Cellulose Thimbles
Velp Scientifica™ Cellulose Thimbles for use with VELP Sovlent Extractors.
Brand:  Velp Scientifica™ A00000295
Product Code. 15551644
  • 121.00€ / Pack of 25
Only null left
Estimated Shipment: 03-07-2025
Description
  • For use with VELP Sovlent Extractors
Specifications
Specifications
25
33 mm
VELP Sovlent Extractors
80 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Velp Scientifica™ Cellulose Thimbles > Size: 33 x 80 mm