Learn More
R&D Systems™ CellXVivo Human M1 Macrophage Differentiation Kit
For the differentiation of human CD14+ monocytes into M1 macrophages
Brand:  R&D Systems™ CDK012
321.75 EUR valid until 2025-07-30
Use promo code "24078" to get your promotional price.
Product Code. 30001735
  • 495.00€ / Each
Ultimate Promo May
Use promo code 24078 at checkout
View Available Promotions
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Cell Differentiation
1 Kit
Certificates
Promotions
Promotions
  • R&D Systems , Tocris
    Empowering Research: The Right Reagents for Every Step
    Promo Code: 24078
    Expires2025-07-30
Product Content Correction
Product Title
R&D Systems™ CellXVivo Human M1 Macrophage Differentiation Kit > 1 Kit