Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
49.04€
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
17329822
|
Thermo Scientific™
60106-CH-15 |
Ceramic Homogenizers for 15 mL Tubes, 100/Pk. | 15 mL Tubes |
49.04€
Pack of 100 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
17339822
|
Thermo Scientific™
60106-CH-2 |
Ceramic Homogenizers for 2 mL Tubes, 200/Pk. | 2 mL Tubes |
49.04€
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17349822
|
Thermo Scientific™
60106-CH-50 |
Ceramic Homogenizers for 50 mL Tubes, 100/Pk. | 50 mL Tubes |
49.04€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
Specifications
Ceramic Homogenizer |