missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemetrics™ Ceric Sulfate Titrant with Ferroin Indicator Method Nitrite Titrets Kit
Brand: Chemetrics™ K-7050
This item is not returnable.
View return policy
Specifications
Nitrite, an intermediate in the nitrogen cycle, is formed during the decomposition of organic matter but readily oxidizes to form nitrate. | |
30 | |
500 to 5000ppm | |
Nitrite Titrets Kit |
Cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, titrettor, 25mL sample cup, and instructions | |
1 Each | |
Ceric Sulfate Titrant with Ferroin Indicator |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- nitrite titrets 500-5000mg/l
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash all exposed externalbody areas thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P330-Rinse mouth.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : cerium(IV) sulphate,sulphuric acid, diethylene glycol, ferrous sulphate heptahydrate, 1,10-phenanth
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chemetrics™ Ceric Sulfate Titrant with Ferroin Indicator Method Nitrite Titrets Kit > Range: 500 to 5000ppm
Spot an opportunity for improvement?Share a Content Correction