missing translation for 'onlineSavingsMsg'
Learn More

Paragon Scientific™ Certified Viscosity Check Oil, 10W30

The viscosity values for the certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for the validating the performance of Cold-Cranking Simulator and Kinematic Viscometers by measuring commercially available formulated engine oils

Brand:  Paragon Scientific™ CVCO10W30-5L


 View more versions of this product

Product Code. 16070192

  • 239.00€ / 5 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Certified in accordance to ISO 17025 / ISO 17034
  • Kinematic Viscosity mm2/s (cSt) in accordance with ASTM D445
  • Density g/mL in accordance with ASTM D4052
  • CCS Dynamic Viscosity mPa.s (cP) in accordance with ASTM D5293
  • Viscosity Index in accordance with ASTM D2270
  • Standard Pack Size 500mL and 5L available
  • Fully traceable to National Standards
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications

Specifications

ISO 17025 / ISO 17034
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Paragon Scientific™ Certified Viscosity Check Oil, 10W30 > Quantity: 5L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Nominal values quoted. Certified Values will vary from batch to batch.