Learn More
Thermo Scientific™ CETAC™ Autosampler Sample Probes
Choose a Thermo Scientific™ CETAC™ Sample Probe to suit your matrix and improve elemental analysis results with automated sample introduction with CETAC Autosamplers.
POA
Specifications
Type | Autosampler Sample Probe |
---|---|
For Use With (Equipment) | CETAC Autosampler |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13211400
|
Thermo Scientific™
942347003991 |
Sample Probe (0.5mm) |
POA
Each |
N/A | |||||
Description
Customized autosampler accessories ensure more efficient, more reliable analysis with your Thermo Scientific™ CETAC™ autosampler and Trace Elemental Analyzers. Choose the appropriate CETAC Autosampler Sample Probe from this comprehensive range.
CETAC Standard Sample Probe- 0.5mm id
- 0.8mm id
Wider bore sample probe reduces the risk of blockage for aqueous samples that contain particulate matter
- 0.5mm id
- 0.8mm id
- 1.0mm id for aqueous samples
Designed especially for use with oil based samples, to ensure large particles are filtered out and avoid the risk of blockage.
Specifications
Autosampler Sample Probe | |
CETAC Autosampler |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.