missing translation for 'onlineSavingsMsg'
Learn More

CGP 3466B maleate, Tocris Bioscience™

Product Code. 18094192
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GAPDH inhibitor; neuroprotective

CGP 3466B maleate is an orally active glyceraldehyde-3-phosphate dehydrogenase (GAPDH) inhibitor. Exhibits antiapoptotic activity via inhibition of GAPDH nuclear translocation and mitochondrial complex 1-dependent H2O2 release. CGP 3466B inhibits colon cancer proliferation in vitro and displays neuroprotective activity in vivo.

Chemical Identifiers

CAS 200189-97-5
Molecular Formula C23H21NO5
Molecular Weight (g/mol) 391.423
InChI Key SQAZQLMBEHYFJA-BTJKTKAUSA-N
Synonym omigapil maleate, cgp 3466b maleate, unii-3q69bfz4op, 3q69bfz4op, tch 346 maleate, dibenzo b,f oxepin-10-ylmethyl-methyl-prop-2-ynyl-amine maleate, n-dibenz b,f oxepin-10-ylmethyl-n-methyl-n-2-propynyl amine monomaleic acid salt, n-dibenz b,f oxepin-10-ylmethyl-n-methyl-n-2-propynyl amine maleate, maleic acid; methyl 2-oxatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3 8 ,4,6,9,11,13-heptaen-9-ylmethyl prop-2-yn-1-ylamine
PubChem CID 9821821
IUPAC Name N-(benzo[b][1]benzoxepin-5-ylmethyl)-N-methylprop-2-yn-1-amine;(Z)-but-2-enedioic acid
SMILES CN(CC#C)CC1=CC2=CC=CC=C2OC3=CC=CC=C31.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 391.42
Percent Purity >99%
Chemical Name or Material CGP 3466B maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.