missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Human IgG, FITC, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB29103.1mL
This item is not returnable.
View return policy
Specifications
Chicken anti-Human IgG | |
Polyclonal | |
Chicken polyclonal antibody raised against Human IgG. The antibody is conjugated with FITC. | |
In PBS, pH 7.2 (0.02% sodium azide) | |
Purified human IgG (whole protein). | |
1 mL | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
FITC | |
Immunocytochemistry Immunohistochemistry(1:250) The optimal working dilution should be determined by the end user. | |
Chicken | |
Antigen affinity purification | |
Secondary | |
Store at 4°C and avoid from light. For long term storage dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chicken anti-Human IgG, FITC, Polyclonal, Secondary Antibody, Abnova™ > 1mL; FITC
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction