missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard Solution, WTW™
Click to view available options
Quantity:
500 mL
1000 mL
Description
WTW offers a wide variety of adapted and suitable products for calibration, maintenance and testing for all measuring systems. Safe and comparable measured results are based on the traceability to proven standards as well as on the flawless condition of the measuring systems. WTW has the matching calibration and test equipment as well as special cleaning products for all parameters and measuring systems. Buffering solutions for pH measurement but also conductivity standards are the main products, but also master solutions for ISE measurements as well as special products for online systems round off our assortment.
Specifications
Specifications
Concentration | 10g/L |
Quantity | 1000 mL |
Color | Colorless |
Solution Contains | Chloride |
Boiling Point | 100°C |
Solubility | Miscible with water |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction