missing translation for 'onlineSavingsMsg'
Learn More
Chlorophyll, Water Soluble, MP Biomedicals Product Code.: 11462026

Chlorophyll, Water Soluble, MP Biomedicals

Product Code. 11462026 Shop All MP Biomedical Products
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 11462026

Brand: MP Biomedicals 0521022180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1406-65-1
Molecular Formula C55H72MgN4O5
Quantity 100 g
Synonym unii-yf5q9ejc8y, chlorophyll a, chlorophyll-a, yf5q9ejc8y, chlorophyll, magnesium, 2e,7r,11r-3,7,11,15-tetramethyl-2-hexadecenyl 3s,4s,21r-9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--kappan23,kappan24,kappan25,kappan26-, sp-4-2, unii-00wnz48or9 component, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-1-2-amino-2-oxo-ethyl carbamoyl-4-guanidino-butyl pyrro, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-5-amino-1-2-amino-2-oxo-ethyl carbamoyl pentyl pyrrolid, magnesium, 3,7,11,15-tetramethyl-2-hexadecenyl 9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--n23,n24,n25,n26-, sp-4-2-3s-3alpha 2e,7s*,11s* ,4beta,21beta
InChI Key ATNHDLDRLWWWCB-AENOIHSZSA-M
SMILES CCC1=C(C2=NC1=CC3=C(C4=C(C(C(=C4[N-]3)C5=NC(=CC6=NC(=C2)C(=C6C)C=C)C(C5CCC(=O)OCC=C(C)CCCC(C)CCCC(C)CCCC(C)C)C)C(=O)OC)[O-])C)C.[Mg+2]
Molecular Weight (g/mol) 893.509
PubChem CID 16667503
ChEBI CHEBI:18230
Chemical Name or Material Chlorophyll, Water Soluble
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorophyll, Water Soluble, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.