missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cholecystokinin, CCK (27-33)

For cellular and molecular biology applications

46.71€ - 66.13€

Specifications

Purification Method HPLC
Common Name CCK Heptapeptide
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 948.12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15448509
View Documents
Thermo Scientific Chemicals
J66940.|D
0.5 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
15458509
View Documents
Thermo Scientific Chemicals
J66940.M^
1 mg
66.13€
1mg
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
-30°C to -10°C
948.12
RUO - research use only
CCK Heptapeptide
Solid
Cholecystokinin, CCK (27-33)
Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only