Learn More
Tocris Bioscience™ CHPG Sodium salt
Selective mGlu5 agonist. Sodium salt of CHPG (Cat. No. 1049)
Brand:  Tocris Bioscience™ 3695/50
594.94 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18004302
  • 758.00€ / 50mg
Estimated Shipment: 03-01-2025
Description
CHPG Sodium salt is a sodium salt of the selective mGlu5 agonist CHPG (Cat. No. 1049).
Specifications
Specifications
>99%
CHPG Sodium salt
CHPG Sodium salt
50 mg
223.59