missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromabond™ Drug Polypropylene SPE Column
Brand: Macherey-Nagel™ 730684
This item is not returnable.
View return policy
Specifications
Silica | |
SPE | |
SPE Enrichment of Drugs From Urine or Plasma | |
Chromabond Drug | |
60 Å |
3 mL | |
500 mg | |
45 μm | |
Solid Phase Extraction Column | |
500 m2/g |