Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Adapter
Brand:  Macherey-Nagel™ 730385
Product Code. 10023041
  • 126.00€ / Pack of 10
Estimated Shipment: 20-06-2025
Specifications
15, 45, 70 mL
Adapter
Polyethylene
10 Pack
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Polyethylene Adapter > For Use With: For 15, 45, 70mL