Learn More
Macherey-Nagel™ Chromabond™ PVDF Adapter
Brand:  Macherey-Nagel™ 730105
Product Code. 10072941
  • 201.00€ / Pack of 10
Estimated Shipment: 16-07-2025
Specifications
3 and 6 mL Glass Column
Adapter
PVDF
10 Pack
Product Title
Macherey-Nagel™ Chromabond™ PVDF Adapter > For Use With: For 3 and 6mL Glass Column