missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Chromalite™ HPLC Column for RP and IEX Chromatography
Prepacked stainless steel columns with robust Chromalite™ synthetic polymers for analytical reverse-phase (RP-HPLC) or ion exchange high pressure liquid chromatography (IEXHPLC).
568.24€ - 944.21€
Specifications
pH | 1 to 14 pH |
---|---|
Pore Size | 200 to 300 Å |
Surface Area | >500 m2/g |
Content And Storage | 5 years from date of manufacture; Storage (at 2°C to 20°C) |
Material | Stainless Steel |
Product Code | Brand | Packing Material | Dimensions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packing Material | Dimensions | Price | Quantity & Availability | ||||
17009643
|
Purolite™
LS06841-602 |
Chromalite™ 10AD2S | 4.6 mm x 250 mm | N/A | |||||
17029663
|
Purolite™
LS06950-601 |
Chromalite™ 5AD2 | 4.6 mm x 150 mm | N/A | |||||
17039663
|
Purolite™
LS06950-602 |
Chromalite™ 5AD2 | 4.6 mm x 250 mm | N/A | |||||
17049643
|
Purolite™
LS06842-601 |
Chromalite™ 10AD2Q | 4.6 mm x 150 mm | N/A | |||||
17059643
|
Purolite™
LS06842-602 |
Chromalite™ 10AD2Q | 4.6 mm x 250 mm | N/A | |||||
17079633
|
Purolite™
LS06840-601 |
Chromalite™ 10AD2 | 4.6 mm x 150 mm | N/A | |||||
17089633
|
Purolite™
LS06840-602 |
Chromalite™ 10AD2 | 4.6 mm x 250 mm | N/A | |||||
17099633
|
Purolite™
LS06841-601 |
Chromalite™ 10AD2S | 4.6 mm x 150 mm | N/A | |||||
Description
Analytical Reverse Phase chromatography (RP-HPLC) and ion-exchange chromatography (IEX-HPLC) resins for separation of proteins, peptides and oligonucleotides, and ideal for biomolecule purification and polishing.
- Uniform particle sizes
- Exceptional batch-to-batch consistency
- Simple scale-up from laboratory analyses through to full-scale plant operation
- Highly hydrophobic
- High chemical stability
- More robust than silica materials
- Pressure stability up to 50 bar
- Efficient regeneration
- Pre-packed in stainless steel HPLC columns
- Compliant with FDA regulation 21 CFR 173.65
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
1 to 14 pH | |
>500 m2/g | |
Stainless Steel |
200 to 300 Å | |
5 years from date of manufacture; Storage (at 2°C to 20°C) |