Learn More
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL
Product Code. 11410181
500 mL, 500mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
MDL Number MFCD00010944
Specifications
Physical Form Liquid
Concentration Cr(NO3)3 in 5% HNO3
UN Number UN1789
Quantity 500 mL
Grade Specpure™
Chemical Name or Material Chromium plasma standard solution, Cr 1000μg/mL
Product Title
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL >