Learn More
Chromium solution for ICP 10g/L HCl matrix, Fisher chemical™ Product Code.: 12996274
compliance-icons
Chromium solution for ICP 10g/L HCl matrix, Fisher chemical™
BIOPHARMA
Product Code. 12996274 Shop All Fisher Chemical Products
100 mL, Bottle
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Bottle
Product Code. 12996274
Brand: Fisher Chemical S 213 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Analyte or Component Names Cr
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Cr
Solvent or Matrix HCl
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1112578 Certificate TypeCertificate of Analysis Date12/05/2025 Catalog NumberS 213 100
Lot Number1010630 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 213 100
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Chromium (Cr) 10000mg/l in HCl 5%
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Chromium solution for ICP 10g/L HCl matrix, Fisher chemical™ >