Learn More
Thermo Scientific™ Cimarec™ 10M Controllers
Adjust speed from 100 to 1000rpm with this external control unit required for use with the Mobil 10 and Mobil 25 stirrers.
Brand: Thermo Scientific™ 50119120
This item is not returnable.
View return policy
Description
Adjust speed from 100 to 1000rpm with the Thermo Scientific™ Cimarec™ 10M Controller, an external control unit that is required for use with the Mobil 10 and Mobil 25 stirrers.
- Automatic start-up for safe stirrer acceleration
Specifications
Mobil 10 and Mobil 25 Stirrer | |
Stirrer Controller | |
230 V |
100 to 1000 rpm | |
230 V | |
100 RPM to 1000 RPM |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Thermo Scientific™ Cimarec™ 10M Controllers > 230V EU