missing translation for 'onlineSavingsMsg'
Learn More
Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals Product Code.: 11475473

Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals

Product Code. 11475473
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11475473

Brand: Thermo Scientific Alfa Aesar A13429.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-79-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00008033
InChI Key APEJMQOBVMLION-VOTSOKGWSA-N
Synonym cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine
PubChem CID 5273472
ChEBI CHEBI:76320
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)C=CC(=O)N

Specifications

Melting Point 145°C to 149°C
Quantity 25 g
Beilstein 2040577
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material Cinnamamide, predominantly trans

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10222065 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberA13429.06,A13429.14,A13429.22
Lot NumberG9298A Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13429.06,A13429.14,A13429.22
Lot Number10205583 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13429.06,A13429.14,A13429.22
Lot Number10176763 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13429.06,A13429.14,A13429.22
Lot Number10165511 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13429.06,A13429.14,A13429.22
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash thoroughly after handling.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.