missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99% Product Code.: 10137853

Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99%

Product Code. 10137853
10 g, Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10137853

Brand: Thermo Scientific Chemicals 204910100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2584-71-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00005252
InChI Key PMMYEEVYMWASQN-QWWZWVQMSA-N
Synonym cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acidShow MoreShow Less
PubChem CID 440014
ChEBI CHEBI:16231
IUPAC Name (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CN[C@H](C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Beilstein 22, I, 546
Specific Rotation +56° to +60° (24°C, 589nm) (c=2, water)
Merck Index 10, 4762
Specific Rotation Condition +58° (24°C c=2,water)
Quantity 10 g
Formula Weight 131.13
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material cis-4-Hydroxy-D-proline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0429793 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog Number204910100,204912500,204910010
Lot NumberA0425338 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number204910100,204912500,204910010
Lot NumberA0315672 Certificate TypeCertificate of Analysis Date24/01/2022 Catalog Number204910100,204912500,204910010
Lot NumberA0293424 Certificate TypeCertificate of Analysis Date18/08/2020 Catalog Number204910100,204912500,204910010
Product Identifier
  • cis-4-Hydroxy-D-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.