missing translation for 'onlineSavingsMsg'
Learn More

Citalopram hydrobromide, Tocris Bioscience™

Product Code. 18064492
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent and selective 5-HT uptake inhibitor

Citalopram hydrobromide is a highly selective and potent 5-HT uptake inhibitor with no effect on noradrenalin or dopamine uptake (IC50 values are 1.8, 8800 and 41000 nM respectively). Has negligible activity at a wide range of receptors.

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H22BrFN2O
Molecular Weight (g/mol) 405.311
InChI Key WIHMBLDNRMIGDW-UHFFFAOYSA-N
Synonym citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
PubChem CID 77995
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br

Specifications

Quantity 50 mg
Formula Weight 405.31
Percent Purity >99%
Chemical Name or Material Citalopram hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.