Learn More
Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1 Product Code.: 15858244
Estimated Shipment: 31-07-2025
Only null left
Add to basket
Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1
Product Code. 15858244
Medium, Pack of 12
Click to view available options
Size:
Large
Medium
Small
Unit Size:
Each
Pack of 12
Product Code. 15858244
Brand: Honeywell™ N65550032
Only null left
Estimated Shipment: 31-07-2025
Add to basket
Only null left
Add to basket
High-Performance, Dual-Cartridge (Class 1) Half-Mask Respirator
  • Lightweight and low-profile ergonomic design encourages high levels of user acceptance
  • Unique cradle head band and yoke piece for ease of adjustment
  • Contoured revert seal ensures an excellent face seal that will not cause discomfort
  • Thermoplastic elastomer material avoids latex allergens that are common with economy masks
  • Single unit, four-point harness with neck strap is easy to use
  • Harness's ergonomic design and improved suspension system fits perfectly on top of the head and offers a high level of comfort
  • Superior exhalation valve allows more breath to be exhaled makes workers feel cooler and more comfortable
  • Reduced dead air space means less re-breathed exhaled air
  • Variable thickness in sealing area conforms to nose bridge
  • Filter cartridges not included, available for order separately
Order Info
Filter cartridges not included, available for order separately
TRUSTED_SUSTAINABILITY
Specifications
Cartridge/Filter Filter Cartridges Sold Separately
Cartridge Type Dual Cartridge: Class 1
Certifications/Compliance EC Category PPE: 3, EC Certificate: 251, EC Attestation: CE Ceert 251-5500 Respir-Dec 2007, EN 140, Declaration of conformity INSPEC N65550031
Color Gray
Material Thermoplastic Elastomer
Size Medium
For Use With (Application) Chemical, Laboratory, and Petrochemical Industries
Includes 4-Point Head Harness
Product Content Correction
Product Title
Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1 >