Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
Brand: Hirschmann™ 1300170
This item is not returnable.
View return policy
Description
- One mark
- Amber stain graduation
- Acc. to DIN EN ISO 648
Specifications
B | |
Yes | |
6/Pk. | |
25 mL | |
Volumetric Pipet |
Glass | |
Brown | |
0.045 mL | |
530 mm |
Product Content Correction
Product Title
Hirschmann™ AR™ Glass Volumetric Pipettes > Capacity: 25mL