Learn More
Zwiebel™ Class E2 Weight with Certificate
Zwiebel™ E2 Weight with Certificate is intended for use in the verification or calibration of class F1 weights.
Brand:  Zwiebel™ PBI2302L
Product Code. 11750633
  • 201.00€ / Each
Estimated Shipment: 24-03-2025
Description
  • Permanent magnetization: < 8 μT
  • Weights intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I. Class E2 weights or weight sets shall be accompanied by a calibration certificate
  • One bloc material
Specifications
Specifications
Wire Weight
8500 ±100 kg/m3
4.2 mm short sides, 6.3 mm long side
2 mg
Square
Certificate
Product Title
Zwiebel™ Class E2 Weight with Certificate > Weight: 2 mg