Learn More
Tenak™ Classic Rack for Chest Freezers, 50 mm Box Height
Classic Rack is suitable for use with all chest freezer models.
72.00€ - 82.15€
Specifications
Includes | Locking rod |
---|---|
Holds | Boxes |
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 50 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11517244
|
Tenak™
TE21440 |
Boxes |
82.15€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
11557244
|
Tenak™
TE21450 |
Boxes |
75.55€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
11567244
|
Tenak™
TE21451 |
Boxes |
79.15€
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15220484
|
Tenak™
TE21448 |
Boxes |
72.00€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
15240484
|
Tenak™
TE21449 |
Boxes |
74.40€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
50 mm (Box) |
Boxes | |
Chest Frseezers |