Learn More
Abeba™ Clean Room 3620 Shoes
Special ESD clean room shoes.
Brand:  Abeba™ 3620/42
87.85 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15670378
  • 109.00€ / 1 pair
Estimated Shipment: 21-01-2025
Description
  • CE EN ISO 20347:2012 OB A, E SRA
  • Microfiber
  • White
  • ESD dissipative according to DIN EN 61340
  • Fixed gaitor, cleanroom textile
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRA
Microfiber
3620
White
42