Learn More
Zeiss™ Clear Glass Plate
Clear glass plate for use with Zeiss Microscopes
Brand:  Zeiss™ 4355010002
Product Code. 11805133
  • 69.05€ / Each
Estimated Shipment: 20-06-2025
Description
Compatible Microscopes:
  • Axio Zoom.V16
  • SteREO Discovery.V12
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Specifications
Plate
120 mm
Glass
1 Ea.
Documents
One moment while we fetch your results.
Product Title
Zeiss™ Clear Glass Plate > Diameter: 120mm