Learn More
Nickel Electro™ Clifton™ Stainless Steel Raised Shelf
Obtain a good even heat distibution with perforted shelves. Nickel Electro™ Clifton™ Stainless Steel Raised Shelf allows two immersion depths by inverting the shelf.
123.00€ - 320.00€
Product Code | Brand | Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
12748898
|
Nickel Electro™
RS4-14 |
30 x 30 x 20 cm | 14, 22, 28 And 38 Liter Baths |
123.00€
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
12768898
|
Nickel Electro™
RS4-38 |
60 x 35 x 20 cm | 38 Liter Baths |
320.00€
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
12758898
|
Nickel Electro™
RS4-22 |
30 x 35 x 20 cm | NE4-22 T, NE4-28 T, NE4-28 D, NE4-22 E Stirred Thermostatic Water Baths |
151.00€
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
Description
- Simple solution for placing shallow vessels in a stirred bath
- Manufactured from stainless steel
- The raised shelf is perforated to allow free circulation of water
- Can be easily removed and cleaned
- Shelf can be inverted to provide two different immersion depths
Specifications
Shelf Raised | |
Stainless Steel |