Learn More
Bollé Safety COBRA Industrial Spare Lens, Clear Product Code.: 12609190
Bollé Safety COBRA Industrial Spare Lens, Clear
Product Code. 12609190
Product Code. 12609190
Brand: Bollé Safety OCCOBPSI
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with COBRA industrial Safety Glasses
  • Bollé Safety clear lenses provide 90% light transmission
  • Is compatible with the PLATINUM anti-fog coating.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Thickness (Metric) 2.2 mm
    Lens Coating Anti-fog, Scratch-resistant
    Disposable Reusable
    Lens Material Polycarbonate
    Sterility Non-sterile
    • Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Bolle Safety Cobra
      Product Content Correction
      Product Title
      Bollé Safety COBRA Industrial Spare Lens, Clear >