Learn More
COD Standard, 200mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Concentration | 200 mg/L |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
COD20025B1 | Certificate of Analysis | 15/04/2025 | COD200 |
COD20023J1 | Certificate of Analysis | 05/12/2023 | COD200 |
COD20023F1 | Certificate of Analysis | 17/08/2023 | COD200 |
COD20022G1 | Certificate of Analysis | 08/12/2022 | COD200 |
COD20022B1 | Certificate of Analysis | 30/03/2022 | COD200 |
Product Content Correction
Product Title
COD Standard, 200mg/L, Reagecon™ >