Learn More
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate
Two matched 1 mg/mL vials (100 ÎĽg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
Brand:  Novus Biologicals™ NBP2-47073-2VIAL
Product Code. 18196748
  • 655.00€ / Pack of 2
Description
The Colon Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 Vials
Store at -80C. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate > 2 Vial