Learn More
Shamrock Color Coded Multi-Purpose Labeling Tape
This Color Coded Multi-Purpose Labeling Tape is designed for organization in laboratories and is removable.
Brand: Shamrock ST-12-16
Description
- Removable labeling tape for organization in laboratories
- Assorted colors and sizes available
Specifications
Removable | |
Roll | |
500 in. | |
1 in. (Core) |
Aqua | |
Colored Label Tape | |
0.5 in. |