missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CometAssay LMAgarose
1 bottle (100 mL) of a CometAssay™ LMAgarose
Brand: R&D Systems™ 4250-500-02
This item is not returnable.
View return policy
Description
- Store bottle at 2-8 °C
- CometAssay™ LMAgarose is ready to use once molten
- Loosen the cap to allow for expansion then heat the bottle in a 90-100 °C water bath for 5 minutes, or until the agarose is molten (microwaving is not recommended)
- Place the bottle in a 37 °C water bath for at least 20 minutes to cool
- The LMAgarose will remain molten at 37 °C for sample preparation indefinitely
- CometAssay™ LMAgarose is 1% low melting agarose in PBS
- Designed for use with CometAssay™, FLARE™, and CometChip™ Kits

Specifications
CometAssay™, FLARE™, and CometChip™ Kits | |
100 mL | |
Store the unopened product at 2 - 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ CometAssay LMAgarose > Quantity: 100 mL
Spot an opportunity for improvement?Share a Content Correction