Learn More
Hamilton™ Communication Cable from PC to Microlab 600 RS-232 Port
Brand:  Hamilton™ 3553-01
Product Code. 15349712
  • 117.00€ / Each
Only null left
Estimated Shipment: 27-06-2025
Specifications
DB-9 Female upstream and DB-9 Male downstream
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hamilton™ Communication Cable from PC to Microlab 600 RS-232 Port >