Learn More
Nabertherm™ Compact Tube Furnace with B510 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
5960.00€ - 12170.00€
Product Code | Brand | Diameter (Metric) Process Tube | Heated Zone | Temperature (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Process Tube | Heated Zone | Temperature (Metric) | Price | Quantity & Availability | ||||
17318089
|
Nabertherm™
R-322L2AN |
120 mm | 500 mm | 1200°C |
5960.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17328089
|
Nabertherm™
R-322L3AN |
120 mm | 500 mm | 1300°C |
6765.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17388089
|
Nabertherm™
R-472L3AN |
100 mm | 750 mm | 1300°C |
10230.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17378089
|
Nabertherm™
R-472L2AN |
170 mm | 750 mm | 1200°C |
9045.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17338099
|
Nabertherm™
R-572L2AN |
170 mm | 1000 mm | 1200°C |
10675.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17348099
|
Nabertherm™
R-572L3AN |
170 mm | 1000 mm | 1300°C |
12170.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
Description
- Excellent price - performance ratio
- Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
- Temperature, max. 1200°C or 1300°C
- Single-zoned design as standard with controller B 510
- Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
- Dual shell housing made of sheets of textured stainless steel
- Insulation made of non-classified fibre material
Specifications
B510 | |
Compact Tube Furnace |
Product Title
Nabertherm™ Compact Tube Furnace with B510 Controller