Learn More
Oxoid UK Compact W-Zip Seal Pouches 20pcs
Compact W-Zip Seal Pouches 20pcs with integral seal
Brand:  Oxoid UK AG0060C
80.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10689754
  • 87.55€ / Pack of 20
In Stock
Description
Compact W-Zip Seal Pouches 20pcs with integral seal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Compact Seal Pouch
20/Pk.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6259112 Certificate TypeCertificate of Analysis Date25/04/2025 Catalog NumberAG0060C
Lot Number6235786 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberAG0060C
Lot Number6215243 Certificate TypeCertificate of Analysis Date25/12/2024 Catalog NumberAG0060C
Lot Number6191451 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberAG0060C
Lot Number6180683 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberAG0060C
Promotions
Promotions
Product Content Correction
Product Title
Oxoid UK Compact W-Zip Seal Pouches 20pcs > Compact W-Zip Seal Pouches 20pcs