missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complex III Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5144114
This item is not returnable.
View return policy
Description
Complex III, or ubiquinol cytochrome c oxidoreductase, is a complex of 11 subunits and catalyses electron transfer from reduced ubiquinone or coenzyme Q 10 to cytochrome c. The complex contains 11 different subunits, one of which, cytochrome b, is encoded on mitochondrial DNA, while the remainder subunits are nuclear-encoded. Genetic complex III deficiencies occur but are rare.
Specifications
Complex III | |
Polyclonal | |
Unconjugated | |
UQCRC2 | |
0710008D09Rik, 1100001F06Rik, 1110020P15Rik, 1110032G10Rik, 1500004O06Rik, 1500040F11Rik, 2210415M14Rik, 2210416J04Rik, 2310021J10Rik, 2610002H19Rik, 2610041P16Rik, 4430402G14Rik, 5830407L17Rik, 8 kDa subunit 9, 9.5kDa, AA408921, AA959903, AA960494, AI875505, AL022707, c1502, COB, Complex III subunit 1, complex III subunit 10, Complex III subunit 2, Complex III subunit 3, Complex III subunit 4, complex III subunit 5, Complex III subunit 6, complex III subunit 7, Complex III subunit 8, complex III subunit 9, Complex III subunit III, Complex III subunit IV, Complex III subunit IX, Complex III subunit VII, complex III subunit VIII, Complex III subunit X, complex III subunit XI, Core protein I, core protein II, CYC1, CYTB, Cytochrome b, cytochrome bc-1 complex core protein II, Cytochrome b-c1 complex subunit 1, mitochondrial, cytochrome b-c1 complex subunit 10, Cytochrome b-c1 complex subunit 11, Cytochrome b-c1 complex subunit 2, mitochondrial, Cytochrome b-c1 complex subunit 3, Cytochrome b-c1 complex subunit 4, cytochrome b-c1 complex subunit 5, Cytochrome b-c1 complex subunit 6, mitochondrial, cytochrome b-c1 complex subunit 7, Cytochrome b-c1 complex subunit 8, cytochrome b-c1 complex subunit 9, Cytochrome b-c1 complex subunit Rieske, mitochondrial, cytochrome c1, cytochrome c-1, cytochrome c1 non-heme 11 kDa protein, Cytochrome c1 non-heme 7 kDa protein, cytochrome c1, heme protein, mitochondrial, cytochrome C1, nonheme 11 kDa protein, cytochrome C1, nonheme 7kDa protein, D3S3191, HSPC051, HSPC119, HSPC151, hypothetical protein LOC613899, hypothetical protein LOC616871, liver regeneration-related protein LRRGT00195, Low molecular mass ubiquinone-binding protein, low molecular mass ubiquinone-binding protein (9.5kD), low molecular mass ubiquinone-binding protein (Qpc-pending), LRRGT00195, MC3DN3, MC3DN4, MC3DN5, MC3DN6, Mitochondrial hinge protein, mitochondrial ubiquinone-binding protein, mitochondrially encoded cytochrome b, MTCYB, MT-CYB, QCR1, QCR10, QCR2, QCR6, QCR7, QCR8, QCR9, QPC, QP-C, Rieske iron-sulfur, Rieske iron-sulfur protein, Rieske iron-sulfur protein precursor, Rieske protein UQCRFS1, RIP1, RIS1, RISP, similar to ubiquinol-cytrochrome-c reductase (subunit II), smallest subunit of ubiquinol-cytochrome c reductase, Su9, ubiquinol cytochrome c reductase core protein 2, ubiquinol cytochrome c reductase hinge protein, ubiquinol cytochrome c reductase, 6.4kDa subunit, ubiquinol--cytochrome c reductase, ubiquinol-cytochrome c reductase (6.4kD) subunit, Ubiquinol-cytochrome c reductase 8 kDa protein, ubiquinol-cytochrome c reductase binding protein, ubiquinol-cytochrome c reductase binding protein-like, ubiquinol-cytochrome c reductase complex (7.2 kD), ubiquinol-cytochrome c reductase complex 11 kDa protein, Ubiquinol-cytochrome c reductase complex 14 kDa protein, ubiquinol-cytochrome c reductase complex 6.4 kDa protein, Ubiquinol-cytochrome c reductase complex 7.2 kDa protein, ubiquinol-cytochrome c reductase complex 7.2kDa protein, ubiquinol-cytochrome c reductase complex 9.5 kDa protein, ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II), ubiquinol-cytochrome c reductase complex III subunit VII, ubiquinol-cytochrome c reductase complex protein, ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C, ubiquinol-cytochrome c reductase core protein 1, ubiquinol-cytochrome c reductase core protein 2, ubiquinol-cytochrome c reductase core protein I, ubiquinol-cytochrome c reductase core protein II, ubiquinol-cytochrome c reductase hinge protein, ubiquinol-cytochrome c reductase iron-sulfur subunit, ubiquinol-cytochrome c reductase subunit, ubiquinol-cytochrome c reductase, 6.4kDa subunit, ubiquinol-cytochrome c reductase, complex III subunit VI, ubiquinol-cytochrome c reductase, complex III subunit VII, ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa, ubiquinol-cytochrome c reductase, complex III subunit VIII | |
Rabbit | |
Protein G | |
RUO | |
10975, 1537, 27089, 29796, 4519, 7381, 7384, 7385, 7386, 7388 | |
-20°C or -80°C if preferred | |
IgG |
ELISA, Immunocytochemistry | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
O14949, O14957, P00156, P07919, P08574, P14927, P22695, P31930, P47985, Q9UDW1 | |
CYC1, MT-CYB, UQCR10, UQCR11, UQCRB, UQCRC1, UQCRC2, UQCRFS1, UQCRH, UQCRQ | |
Recombinant Human Cytochrome b-c1 complex subunit 10 protein (1-56AA) | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Complex III Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction