Learn More
84 μS/cm Conductivity Standards, Hanna Instruments™
Click to view available options
Quantity:
500 mL
Description
- Calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Premium quality 84 μS/cm conductivity solution standardized with a conductivity meter
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
- Air tight bottle with tamper-proof seal to ensure quality
Specifications
Specifications
Packaging | Bottles |
Quantity | 500 mL |
Product Content Correction
Product Title
84 μS/cm Conductivity Standards, Hanna Instruments™ >