Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand: Fisherbrand™ 9001041-010B
Description
- Connecting Cable with BNC Plug
- For combined ion-sensitive electrode with solid membranes
- For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
BNC | |
Connecting Cable | |
For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug . | |
3 m |
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31